Assay Details
| Gene Symbol: Erc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177814.4 | NP_808482.2 | 12 - 13 | 2707 | 61 | |
| GenBank mRNA | AK051416.1 | - | 2 - 3 | 199 | 61 | |
| AK122265.1 | - | 11 - 12 | 1431 | 61 | ||
| AK147213.1 | - | 15 - 16 | 2794 | 61 | ||
| AY356530.1 | - | 13 - 14 | 2442 | 61 | ||
| AY356531.1 | - | 11 - 12 | 2394 | 61 | ||
| AY356532.1 | - | 12 - 13 | 2406 | 61 | ||
| AY356533.1 | - | 11 - 12 | 2394 | 61 | ||
| BC056760.1 | - | 12 - 13 | 2707 | 6331997 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erc2
Genomic Map