Assay Details
| Gene Symbol: Ccdc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177111.3 | NP_796085.3 | 8 - 9 | 1084 | 67 | |
| GenBank mRNA | AK145297.1 | - | 7 - 8 | 1097 | 67 | |
| AK158666.1 | - | 6 - 7 | 602 | 67 | ||
| BC070471.1 | - | 8 - 9 | 1084 | 30626499 | 67 | |
| BC072583.1 | - | 79 | 6403859 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158666.1, BC072583.1 transcripts.
Set Membership: |
|

Ccdc66
Genomic Map