Assay Details
| Gene Symbol: Ppp1cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172707.3 | NP_766295.2 | 3 - 4 | 796 | 151 | |
| GenBank mRNA | AK004686.2 | - | 3 - 4 | 507 | 151 | |
| AK088893.1 | - | 3 - 4 | 436 | 151 | ||
| AK089067.1 | - | 615 | 151 | |||
| AK147112.1 | - | 3 - 4 | 599 | 151 | ||
| AK160743.1 | - | 3 - 4 | 505 | 151 | ||
| AK166168.1 | - | 3 - 4 | 714 | 151 | ||
| AK166416.1 | - | 3 - 4 | 800 | 151 | ||
| AK166623.1 | - | 3 - 4 | 699 | 151 | ||
| AK168141.1 | - | 3 - 4 | 693 | 151 | ||
| AK168512.1 | - | 3 - 4 | 442 | 151 | ||
| AK169379.1 | - | 3 - 4 | 545 | 151 | ||
| AK171283.1 | - | 3 - 4 | 665 | 151 | ||
| AK193890.1 | - | 1 - 2 | 70 | 151 | ||
| AK215769.1 | - | 1 - 2 | 70 | 151 | ||
| BC046832.1 | - | 3 - 4 | 497 | 6812937 | 151 | |
| M27073.1 | - | 3 - 4 | 505 | 151 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ppp1cb
Genomic Map