Assay Details
Gene Symbol: Rbks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153196.1 | NP_694876.1 | 2 - 3 | 238 | 116 | |
GenBank mRNA | AK017229.1 | - | 2 - 3 | 256 | 116 | |
AK138939.1 | - | 2 - 3 | 256 | 116 | ||
BC023339.1 | - | 2 - 3 | 238 | 5253827 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
