Assay Details
Gene Symbol: Gm5886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177652.2 | NP_001171123.1 | 1 - 2 | 99 | 84 | |
NR_111985.1 | 1 - 2 | 99 | 84 | |||
GenBank mRNA | AB256662.1 | - | 1 - 2 | 86 | 84 | |
BC055452.1 | - | 1 - 2 | 101 | 84 | ||
BC055924.1 | - | 1 - 2 | 87 | 84 | ||
BC069936.1 | - | 1 - 2 | 87 | 84 | ||
BC087541.1 | - | 1 - 2 | 94 | 84 | ||
BC108376.1 | - | 1 - 2 | 93 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
