Assay Details
Gene Symbol: Ppp2r2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172994.2 | NP_766582.1 | 5 - 6 | 877 | 58 | |
GenBank mRNA | AK032023.1 | - | 5 - 6 | 780 | 58 | |
AK049830.1 | - | 5 - 6 | 763 | 58 | ||
AY348866.1 | - | 5 - 6 | 879 | 58 | ||
BC059811.1 | - | 5 - 6 | 877 | 6849535 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
