Assay Details
| Gene Symbol: Sis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081137.1 | NP_001074606.1 | 15 - 16 | 1861 | 78 | |
| GenBank mRNA | EU937530.1 | - | 16 - 17 | 1924 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sis
Genomic Map