Assay Details
| Gene Symbol: Sis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081137.1 | NP_001074606.1 | 22 - 23 | 2530 | 136 | |
| GenBank mRNA | EU937530.1 | - | 23 - 24 | 2593 | 136 | |
More Information
Set Membership: |
|

Sis
Genomic Map