Assay Details
| Gene Symbol: Afap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027373.2 | NP_081649.1 | 14 - 15 | 2035 | 74 | |
| GenBank mRNA | AK129483.1 | - | 14 - 15 | 1962 | 74 | |
| AK147706.1 | - | 14 - 15 | 1927 | 74 | ||
| AK148000.1 | - | 14 - 15 | 1890 | 74 | ||
| AK161385.1 | - | 14 - 15 | 1986 | 74 | ||
| BC050814.1 | - | 14 - 15 | 2035 | 6773872 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afap1
Genomic Map