Assay Details
| Gene Symbol: Afap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027373.2 | NP_081649.1 | 1 - 2 | 222 | 67 | |
| GenBank mRNA | AK011153.1 | - | 1 - 2 | 50 | 67 | |
| AK080756.1 | - | 1 - 2 | 157 | 67 | ||
| AK129483.1 | - | 1 - 2 | 149 | 67 | ||
| AK147706.1 | - | 1 - 2 | 114 | 67 | ||
| AK148000.1 | - | 1 - 2 | 77 | 67 | ||
| BC050814.1 | - | 1 - 2 | 222 | 6773872 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011153.1, AK080756.1, AK147706.1, AK148000.1, BC050814.1, NM_027373.2 transcripts.
Set Membership: |
|

Afap1
Genomic Map