Assay Details
| Gene Symbol: Afap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027373.2 | NP_081649.1 | 4 - 5 | 550 | 65 | |
| GenBank mRNA | AK011153.1 | - | 4 - 5 | 378 | 65 | |
| AK080756.1 | - | 4 - 5 | 485 | 65 | ||
| AK129483.1 | - | 4 - 5 | 477 | 65 | ||
| AK147706.1 | - | 4 - 5 | 442 | 65 | ||
| AK148000.1 | - | 4 - 5 | 405 | 65 | ||
| AK161385.1 | - | 4 - 5 | 501 | 65 | ||
| BC050814.1 | - | 4 - 5 | 550 | 6773872 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afap1
Genomic Map