Assay Details
| Gene Symbol: Ift80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026641.2 | NP_080917.1 | 18 - 19 | 2286 | 106 | |
| GenBank mRNA | AK019542.1 | - | 5 - 6 | 2203 | 106 | |
| AK042231.1 | - | 18 - 19 | 2211 | 106 | ||
| AK147305.1 | - | 17 - 18 | 5813 | 106 | ||
| BC013814.1 | - | 13 - 14 | 1575 | 106 | ||
| BC034101.1 | - | 18 - 19 | 2181 | 4501451 | 106 | |
More Information
Set Membership: |
|

Ift80
Genomic Map