Assay Details
| Gene Symbol: Ift80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026641.2 | NP_080917.1 | 9 - 10 | 1148 | 66 | |
| GenBank mRNA | AK042231.1 | - | 9 - 10 | 1073 | 66 | |
| AK129339.1 | - | 9 - 10 | 1076 | 66 | ||
| AK147305.1 | - | 8 - 9 | 4675 | 66 | ||
| BC013814.1 | - | 4 - 5 | 437 | 66 | ||
| BC034101.1 | - | 9 - 10 | 1043 | 4501451 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ift80
Genomic Map