Assay Details
| Gene Symbol: Asb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170748.1 | NP_001164219.1 | 8 - 9 | 1576 | 78 | |
| NM_080856.4 | NP_543132.3 | 8 - 9 | 1576 | 78 | ||
| GenBank mRNA | AF403042.1 | - | 3 - 4 | 993 | 78 | |
| AK052884.1 | - | 8 - 9 | 1578 | 78 | ||
| AK164800.1 | - | 2 - 3 | 639 | 78 | ||
| BC145002.1 | - | 3 - 4 | 962 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301277 | AK164800.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301277 | AF403042.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301277 | NM_001170748.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301277 | AK164800.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301277 | AF403042.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P301277 | NM_001170748.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133965 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702837 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666961 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587309 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 256105 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559262 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 134627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308211 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 505399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409235 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 175918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 3239 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74989 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 42382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119543 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107215 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179529 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301277 | AK164800.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301277 | AF403042.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301277 | NM_001170748.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301277 | AK164800.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301277 | AF403042.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P301277 | NM_001170748.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484061 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29484059 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08061240 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107215 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179529 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08061267 | - | - |
| Whole Mouse Genome G4122A | A_51_P301277 | AK164800.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301277 | AF403042.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301277 | NM_001170748.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301277 | AK164800.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301277 | AF403042.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P301277 | NM_001170748.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120739 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127120 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179529 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107215 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11240511 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11352910 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11425995 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11309806 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11309805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29483958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29484051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483972 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11789897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11816439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483985 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11245992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11777382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11352903 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11274835 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11398624 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11285027 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11352904 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11729096 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11381575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11398632 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11582732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484020 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483968 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11463921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483973 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484048 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11352913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29483994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29484022 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10413390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413385 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413391 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413383 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413389 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413388 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413384 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10413382 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449547_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10413381 | - | - |
| Mouse Genome 430 2.0 Array | 1449547_at | - | - |
| Mouse Genome 430 2.0 Array | 1449547_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444742_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444742_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 142687_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 142687_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400002197.mm.1 | NM_080856.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002197.mm.1 | NM_080856.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002197.mm.1 | TC1400000267.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400002196.mm.1 | NM_001170748.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002196.mm.1 | NM_001170748.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002196.mm.1 | NM_080856.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002196.mm.1 | NM_080856.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400002196.mm.1 | TC1400000267.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449547_at | - | - |
| Mouse Expression Set 430 | 1449547_PM_at | - | - |
| Mouse Expression Set 430 | 1446424_at | - | - |
| Mouse Expression Set 430 | 1446424_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449547_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449547_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102810368 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5860427 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102810368 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2610760 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250663 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2610760 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102810368 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102810368 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250663 | - | - |

Asb14
Genomic Map