Assay Details
| Gene Symbol: Asb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170748.1 | NP_001164219.1 | 9 - 10 | 1726 | 73 | |
| GenBank mRNA | AF403042.1 | - | 4 - 5 | 1143 | 73 | |
| AK164800.1 | - | 3 - 4 | 789 | 73 | ||
| BC145002.1 | - | 4 - 5 | 1112 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Asb14
Genomic Map