Assay Details
| Gene Symbol: Ifnar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010508.2 | NP_034638.2 | 4 - 5 | 650 | 106 | |
| GenBank mRNA | AK132431.1 | - | 4 - 5 | 582 | 106 | |
| AK141630.1 | - | 4 - 5 | 586 | 106 | ||
| AK148246.1 | - | 4 - 5 | 592 | 106 | ||
| AK157555.1 | - | 4 - 5 | 617 | 106 | ||
| BC043935.1 | - | 4 - 5 | 601 | 4507009 | 106 | |
| BC052217.1 | - | 4 - 5 | 610 | 30008497 | 106 | |
| BC052429.1 | - | 4 - 5 | 612 | 6836300 | 106 | |
| M89641.1 | - | 4 - 5 | 615 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifnar1
Genomic Map