Assay Details
| Gene Symbol: Ifnar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010508.2 | NP_034638.2 | 7 - 8 | 1124 | 90 | |
| GenBank mRNA | AK132431.1 | - | 7 - 8 | 1056 | 90 | |
| AK136950.1 | - | 7 - 8 | 1100 | 90 | ||
| AK141630.1 | - | 7 - 8 | 1060 | 90 | ||
| AK148246.1 | - | 7 - 8 | 1066 | 90 | ||
| AK157555.1 | - | 8 - 9 | 1152 | 90 | ||
| BC043935.1 | - | 7 - 8 | 1075 | 4507009 | 90 | |
| BC052429.1 | - | 7 - 8 | 1086 | 6836300 | 90 | |
| M89641.1 | - | 7 - 8 | 1089 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ifnar1
Genomic Map