Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600007674.mm.1 |
NM_001199210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007674.mm.1 |
TC1600000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600007675.mm.1 |
NM_001199210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007675.mm.1 |
NM_027627.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007675.mm.1 |
TC1600000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600007676.mm.1 |
NM_001199210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007676.mm.1 |
NM_027627.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007676.mm.1 |
TC1600000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600007678.mm.1 |
NM_001199210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007678.mm.1 |
NM_027627.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007678.mm.1 |
TC1600000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600007679.mm.1 |
NM_001199210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007679.mm.1 |
NM_027627.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007679.mm.1 |
TC1600000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600007677.mm.1 |
NM_001199210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007677.mm.1 |
NM_027627.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007677.mm.1 |
TC1600000989.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600007673.mm.1 |
NM_001199210.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600007673.mm.1 |
TC1600000989.mm.1 |
MTA TC |