Assay Details
| Gene Symbol: Ddx3y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012008.2 | NP_036138.1 | 2 - 3 | 200 | 116 | |
| GenBank mRNA | AJ007376.1 | - | 2 - 3 | 167 | 116 | |
| BC021453.1 | - | 2 - 3 | 165 | 5043470 | 116 | |
| CD552910.1 | - | 2 - 3 | 200 | 116 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx3y
Genomic Map