Assay Details
| Gene Symbol: Mmp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019724.3 | NP_062698.2 | 2 - 3 | 523 | 68 | |
| GenBank mRNA | AB021228.1 | - | 2 - 3 | 338 | 68 | |
| AF282844.1 | - | 2 - 3 | 290 | 68 | ||
| AK046170.1 | - | 2 - 3 | 382 | 68 | ||
| AK048543.1 | - | 2 - 3 | 403 | 68 | ||
| AK053598.1 | - | 2 - 3 | 413 | 68 | ||
| AK077939.1 | - | 567 | 68 | |||
| BC057926.1 | - | 2 - 3 | 525 | 5292478 | 68 | |
More Information
Set Membership: |
|

Mmp16
Genomic Map