Assay Details
| Gene Symbol: Dnajb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037941.3 | NP_001033030.2 | 6 - 7 | 692 | 71 | |
| GenBank mRNA | AK005680.1 | - | 6 - 7 | 597 | 71 | |
| BF660899.1 | - | 1 - 2 | 71 | |||
| DV660970.1 | - | 5 - 6 | 487 | 71 | ||
More Information
Set Membership: |
|

Dnajb6
Genomic Map