| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500019635.mm.1 |
TC0500002158.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500019635.mm.1 |
NM_028483.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500019635.mm.1 |
NM_028483.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500019637.mm.1 |
TC0500002158.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500019637.mm.1 |
NM_028483.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500019637.mm.1 |
NM_028483.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500019636.mm.1 |
TC0500002158.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500019636.mm.1 |
NM_028483.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500019636.mm.1 |
NM_028483.1 |
Refseq |