Assay Details
| Gene Symbol: C2cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174847.2 | NP_777272.2 | 10 - 11 | 1618 | 105 | |
| GenBank mRNA | AK017896.1 | - | 10 - 11 | 1603 | 105 | |
| AK077731.1 | - | 10 - 11 | 1579 | 105 | ||
| AK080956.1 | - | 10 - 11 | 1562 | 105 | ||
| AK086520.1 | - | 10 - 11 | 1621 | 105 | ||
| AK143875.1 | - | 10 - 11 | 1592 | 105 | ||
| AK169902.1 | - | 10 - 11 | 1554 | 105 | ||
| AK220454.1 | - | 10 - 11 | 1603 | 105 | ||
| BC043688.1 | - | 10 - 11 | 1579 | 5136661 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

C2cd2
Genomic Map