Assay Details
| Gene Symbol: C2cd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174847.2 | NP_777272.2 | 9 - 10 | 1443 | 55 | |
| GenBank mRNA | AK017896.1 | - | 9 - 10 | 1428 | 55 | |
| AK077731.1 | - | 9 - 10 | 1404 | 55 | ||
| AK080956.1 | - | 9 - 10 | 1387 | 55 | ||
| AK086520.1 | - | 9 - 10 | 1446 | 55 | ||
| AK143875.1 | - | 9 - 10 | 1417 | 55 | ||
| AK169902.1 | - | 9 - 10 | 1379 | 55 | ||
| AK220454.1 | - | 9 - 10 | 1428 | 55 | ||
| BC043688.1 | - | 9 - 10 | 1404 | 5136661 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

C2cd2
Genomic Map