Assay Details
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009316.1 | NP_033342.1 | 13 - 14 | 1516 | 80 | |
NM_172688.3 | NP_766276.1 | 12 - 13 | 1435 | 80 | ||
GenBank mRNA | AK046565.1 | - | 12 - 13 | 1401 | 80 | |
AK053911.1 | - | 12 - 13 | 1421 | 80 | ||
AK133642.1 | - | 6 - 7 | 562 | 80 | ||
AK137046.1 | - | 12 - 13 | 1436 | 80 | ||
AK159277.1 | - | 12 - 13 | 1399 | 80 | ||
BC006665.1 | - | 13 - 14 | 1507 | 3499247 | 80 | |
CV559615.1 | - | 6 - 7 | 572 | 80 | ||
D76446.1 | - | 12 - 13 | 1435 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
