| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800015073.mm.1 |
NM_001272057.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800015073.mm.1 |
NM_001272058.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800015073.mm.1 |
NM_001272059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800015073.mm.1 |
NM_007401.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800015073.mm.1 |
TC0800001899.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800015074.mm.1 |
NM_001272057.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800015074.mm.1 |
NM_001272058.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800015074.mm.1 |
NM_001272059.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800015074.mm.1 |
NM_007401.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800015074.mm.1 |
TC0800001899.mm.1 |
MTA TC |