Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800002463.mm.1 |
NM_028000.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002463.mm.1 |
NM_028000.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002463.mm.1 |
TC0800000328.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002460.mm.1 |
NM_028000.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002460.mm.1 |
NM_028000.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002460.mm.1 |
TC0800000328.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002459.mm.1 |
NM_028000.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002459.mm.1 |
NM_028000.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002459.mm.1 |
TC0800000328.mm.1 |
MTA TC |