Assay Details
| Gene Symbol: Kif24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024241.2 | NP_077203.2 | 10 - 11 | 1772 | 77 | |
| GenBank mRNA | AK050683.1 | - | 10 - 11 | 1758 | 77 | |
| AK084967.1 | - | 7 - 8 | 1371 | 77 | ||
| BC067395.1 | - | 10 - 11 | 1764 | 30460089 | 77 | |
More Information
Set Membership: |
|

Kif24
Genomic Map