Assay Details
Gene Symbol: Kif24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024241.2 | NP_077203.2 | 2 - 3 | 777 | 125 | |
GenBank mRNA | AK016916.1 | - | 4 - 5 | 975 | 125 | |
AK050683.1 | - | 2 - 3 | 763 | 125 | ||
AK084967.1 | - | 2 - 3 | 778 | 125 | ||
AK135307.1 | - | 2 - 3 | 778 | 125 | ||
BC067395.1 | - | 2 - 3 | 769 | 30460089 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
