Assay Details
| Gene Symbol: She | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172530.3 | NP_766118.3 | 5 - 6 | 1434 | 85 | |
| GenBank mRNA | AK034662.1 | - | 5 - 6 | 1445 | 85 | |
| AK157580.1 | - | 5 - 6 | 1434 | 85 | ||
| BC109362.1 | - | 6 - 7 | 950 | 40058813 | 85 | |
| BC109363.1 | - | 5 - 6 | 1406 | 40058814 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321903 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321899 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321904 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321901 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321902 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957468 | NM_172530.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833787 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 800101 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 888721 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22759 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 435169 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943187 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431819 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520669 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874678 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45623 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 952608 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 65624 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 881695 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937173 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 296655 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589759 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510452 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940768 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 196812 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 820779 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 158912 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114020 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178596 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115237 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49764 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 2746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 146121 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144467 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P134389 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109329 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957468 | NM_172530.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321899 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321885 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321892 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321918 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321891 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321893 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321914 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321895 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22321903 | - | - |
| Whole Mouse Genome G4122A | A_52_P240834 | AK034662.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240834 | AK157580.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240834 | NM_172530.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128990 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03123563 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03123565 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03043490 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03089529 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321901 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321903 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321898 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321902 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321899 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22321904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22321925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22322020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321873 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321904 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02418315 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321882 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22322031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03043490 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03077234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321892 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02696855 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03189445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321872 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03197975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22322028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02498550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03093679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02928286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22322023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321875 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03093680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03093681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321922 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03189454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03197980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03093686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03197971 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03197973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02898502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02460626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321885 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02733323 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321889 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22321939 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10493605 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493606 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493608 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493609 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493610 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493607 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10493611 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457094_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438227_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10493604 | - | - |
| Mouse Genome 430 2.0 Array | 1457094_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438227_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1438227_at | - | - |
| Mouse Genome 430 2.0 Array | 1457094_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 214547_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 214547_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300005914.mm.1 | NM_172530.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300005914.mm.1 | NM_172530.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300005914.mm.1 | TC0300000772.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300005915.mm.1 | NM_172530.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300005915.mm.1 | NM_172530.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300005915.mm.1 | TC0300000772.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1457094_at | - | - |
| Mouse Expression Set 430 | 1457094_PM_at | - | - |
| Mouse Expression Set 430 | 1438227_PM_at | - | - |
| Mouse Expression Set 430 | 1438227_at | - | - |
| Murine Genome U74v2 Set | 161099_at | - | - |
| Murine Genome U74v2 Set | 161099_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1457094_at | - | - |
| Mouse Genome 430A 2.0 Array | 1457094_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1457094_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1457094_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438227_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438227_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 342435 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6420692 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006420692 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2757019 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2757019 | - | - |
| mouse-6 v1.1 (Illumina) | scl22963.6.1_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6420692 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22963.6.1_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420692 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2757019 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2757019 | - | - |

She
Genomic Map