Assay Details
| Gene Symbol: Noa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019836.3 | NP_062810.1 | 3 - 4 | 1547 | 120 | |
| GenBank mRNA | AB041537.1 | - | 3 - 4 | 1519 | 120 | |
| AK011533.1 | - | 3 - 4 | 1539 | 120 | ||
| AK017793.1 | - | 3 - 4 | 1520 | 120 | ||
| AK019975.1 | - | 3 - 4 | 1529 | 120 | ||
| AK128920.1 | - | 3 - 4 | 1519 | 120 | ||
| AK139955.1 | - | 3 - 4 | 1491 | 120 | ||
| AK157246.1 | - | 3 - 4 | 1547 | 120 | ||
| BC005689.1 | - | 3 - 4 | 600 | 120 | ||
| BC017154.1 | - | 3 - 4 | 1519 | 4910547 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Noa1
Genomic Map