Assay Details
| Gene Symbol: Gsto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010362.2 | NP_034492.1 | 3 - 4 | 468 | 79 | |
| GenBank mRNA | AK027922.1 | - | 3 - 4 | 418 | 79 | |
| AK146834.1 | - | 3 - 4 | 430 | 79 | ||
| AK168383.1 | - | 3 - 4 | 430 | 79 | ||
| BC085165.1 | - | 3 - 4 | 406 | 30359900 | 79 | |
More Information
Set Membership: |
|

Gsto1
Genomic Map