Assay Details
Gene Symbol: Ppat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172146.2 | NP_742158.1 | 10 - 11 | 1854 | 67 | |
GenBank mRNA | AK147750.1 | - | 10 - 11 | 1504 | 67 | |
AK148034.1 | - | 10 - 11 | 1458 | 67 | ||
AK159141.1 | - | 11 - 12 | 2004 | 67 | ||
AK159859.1 | - | 11 - 12 | 2004 | 67 | ||
AK160542.1 | - | 10 - 11 | 1479 | 67 | ||
AK164506.1 | - | 10 - 11 | 1468 | 67 | ||
AK167057.1 | - | 10 - 11 | 1493 | 67 | ||
AK210731.1 | - | 1 - 2 | 137 | 67 | ||
AK216394.1 | - | 1 - 2 | 137 | 67 | ||
BC006053.1 | - | 9 - 10 | 1696 | 67 | ||
BC023841.1 | - | 10 - 11 | 1481 | 5346105 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659258 | BC023841.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659258 | BC006053.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659258 | AK164506.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659258 | AK167057.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659258 | AK148034.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659258 | BC023841.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659258 | BC006053.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659258 | AK164506.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659258 | AK167057.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P659258 | AK148034.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296905 | AK148034.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296905 | AK159141.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296905 | AK159859.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296905 | NM_172146.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296905 | AK017664.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296905 | AK148034.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296905 | AK159141.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296905 | AK159859.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296905 | NM_172146.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P296905 | AK017664.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407661 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768044 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92187 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 41029 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115915 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 943458 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83887 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 531930 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 239144 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677689 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868643 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 841358 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546104 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583524 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 20129 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174035 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 133981 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627104 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384835 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359614 | NM_134236.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471278 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40721 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 2821 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178623 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88339 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153336 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112685 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659258 | BC023841.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659258 | BC006053.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659258 | AK164506.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659258 | AK167057.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659258 | AK148034.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659258 | BC023841.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659258 | BC006053.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659258 | AK164506.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659258 | AK167057.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P659258 | AK148034.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296905 | AK148034.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296905 | AK159141.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296905 | AK159859.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296905 | NM_172146.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296905 | AK017664.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296905 | AK148034.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296905 | AK159141.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296905 | AK159859.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296905 | NM_172146.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P296905 | AK017664.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112685 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05624095 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153336 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05624083 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01139190 | - | - |
Whole Mouse Genome G4122A | A_51_P296905 | AK148034.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296905 | AK159141.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296905 | AK159859.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296905 | NM_172146.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296905 | AK017664.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296905 | AK148034.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296905 | AK159141.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296905 | AK159859.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296905 | NM_172146.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P296905 | AK017664.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659258 | BC023841.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659258 | BC006053.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659258 | AK164506.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659258 | AK167057.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659258 | AK148034.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659258 | BC023841.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659258 | BC006053.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659258 | AK164506.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659258 | AK167057.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P659258 | AK148034.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107828 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112685 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23818924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23818928 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04597429 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04597424 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04597435 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23818920 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10530808 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530811 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530807 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530817 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530814 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530809 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530818 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530810 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530813 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530816 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530812 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10530815 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428543_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452831_PM_s_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1441770_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10530806 | - | - |
Mouse Genome 430 2.0 Array | 1428543_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1452831_s_at | - | - |
Mouse Genome 430 2.0 Array | 1452831_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1441770_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1428543_at | - | - |
Mouse Genome 430 2.0 Array | 1441770_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231327_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 231327_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0500023358.mm.1 | TC0500002642.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500023358.mm.1 | NM_172146.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023358.mm.1 | NM_172146.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023357.mm.1 | TC0500002642.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500023357.mm.1 | NM_172146.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023357.mm.1 | NM_172146.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023359.mm.1 | TC0500002642.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0500023359.mm.1 | NM_172146.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0500023359.mm.1 | NM_172146.2 | Refseq |
Mouse Expression Set 430 | 1428543_PM_at | - | - |
Mouse Expression Set 430 | 1452831_s_at | - | - |
Mouse Expression Set 430 | 1452831_PM_s_at | - | - |
Mouse Expression Set 430 | 1428543_at | - | - |
Mouse Expression Set 430 | 1441770_at | - | - |
Mouse Expression Set 430 | 1441770_PM_at | - | - |
Murine Genome U74v2 Set | 95554_at | - | - |
Murine Genome U74v2 Set | 95553_PM_at | - | - |
Murine Genome U74v2 Set | 95553_at | - | - |
Murine Genome U74v2 Set | 95554_PM_at | - | - |
Murine Genome U74v2 Set | 168502_r_at | - | - |
Murine Genome U74v2 Set | 168502_PM_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1428543_at | - | - |
Mouse Genome 430A 2.0 Array | 1452831_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1452831_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1428543_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428543_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452831_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452831_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428543_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441770_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441770_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 469562 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1980019 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1980019 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 101410519 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001980019 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1245424 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1222130 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1245424 | - | - |
mouse-6 v1.1 (Illumina) | scl26453.11_0-S | - | - |
mouse-6 v1.1 (Illumina) | scl00231327.1_147-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 101410519 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1980019 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00231327.1_147-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980019 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 101410519 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1245424 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1222130 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1245424 | - | - |
