Assay Details
Gene Symbol: Ppat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172146.2 | NP_742158.1 | 5 - 6 | 1148 | 71 | |
GenBank mRNA | AK147750.1 | - | 5 - 6 | 798 | 71 | |
AK148034.1 | - | 5 - 6 | 752 | 71 | ||
AK159141.1 | - | 6 - 7 | 1298 | 71 | ||
AK159859.1 | - | 6 - 7 | 1298 | 71 | ||
AK160542.1 | - | 5 - 6 | 773 | 71 | ||
AK164506.1 | - | 5 - 6 | 762 | 71 | ||
AK167057.1 | - | 5 - 6 | 787 | 71 | ||
BC023841.1 | - | 5 - 6 | 775 | 5346105 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
