Assay Details
| Gene Symbol: Gabra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008066.3 | NP_032092.1 | 7 - 8 | 994 | 127 | |
| GenBank mRNA | AK039055.1 | - | 7 - 8 | 995 | 127 | |
| AK046953.1 | - | 2 - 3 | 1128 | 127 | ||
| AK048165.1 | - | 892 | 127 | |||
| BC115727.1 | - | 7 - 8 | 916 | 40039253 | 127 | |
| M86567.1 | - | 7 - 8 | 745 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gabra2
Genomic Map