Assay Details
| Gene Symbol: Pdgfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083316.1 | NP_001076785.1 | 16 - 17 | 2517 | 67 | |
| NM_011058.2 | NP_035188.2 | 16 - 17 | 2500 | 67 | ||
| GenBank mRNA | AK136490.1 | - | 16 - 17 | 2486 | 67 | |
| AK147267.1 | - | 16 - 17 | 2449 | 67 | ||
| AK163952.1 | - | 16 - 17 | 2564 | 67 | ||
| BC053036.1 | - | 16 - 17 | 2517 | 5704645 | 67 | |
| M84607.1 | - | 16 - 17 | 2510 | 67 | ||
More Information
Set Membership: |
|

Pdgfra
Genomic Map