Assay Details
| Gene Symbol: Pdgfra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083316.1 | NP_001076785.1 | 3 - 4 | 558 | 60 | |
| NM_011058.2 | NP_035188.2 | 3 - 4 | 541 | 60 | ||
| GenBank mRNA | AK035501.1 | - | 3 - 4 | 498 | 60 | |
| AK081664.1 | - | 3 - 4 | 540 | 60 | ||
| AK136490.1 | - | 3 - 4 | 527 | 60 | ||
| AK145097.1 | - | 3 - 4 | 1141 | 60 | ||
| AK147267.1 | - | 3 - 4 | 490 | 60 | ||
| AK163952.1 | - | 3 - 4 | 605 | 60 | ||
| BC020405.1 | - | 3 - 4 | 472 | 60 | ||
| BC037467.1 | - | 3 - 4 | 480 | 60 | ||
| BC053036.1 | - | 3 - 4 | 558 | 5704645 | 60 | |
| M57683.1 | - | 3 - 4 | 440 | 60 | ||
| M84607.1 | - | 3 - 4 | 551 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfra
Genomic Map