Assay Details
| Gene Symbol: Mpv17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294322.1 | NP_001281251.1 | 4 - 5 | 345 | 70 | |
| NM_001294324.1 | NP_001281253.1 | 4 - 5 | 374 | 70 | ||
| NM_008622.6 | NP_032648.1 | 4 - 5 | 345 | 70 | ||
| GenBank mRNA | AK040477.1 | - | 4 - 5 | 315 | 70 | |
| BC013452.1 | - | 3 - 4 | 298 | 4237284 | 70 | |
| CK622051.1 | - | 3 - 4 | 287 | 70 | ||
| DV062332.1 | - | 4 - 5 | 427 | 70 | ||
| M36411.1 | - | 4 - 5 | 327 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mpv17
Genomic Map