Assay Details
| Gene Symbol: Gtf3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027901.2 | NP_082177.2 | 12 - 13 | 2010 | 62 | |
| GenBank mRNA | AK004895.1 | - | 12 - 13 | 1788 | 62 | |
| AK012095.1 | - | 5 - 6 | 607 | 62 | ||
| AK043944.1 | - | 12 - 13 | 1777 | 62 | ||
| AK083585.1 | - | 12 - 13 | 2012 | 62 | ||
| AK083636.1 | - | 11 - 12 | 1765 | 62 | ||
| AK129033.1 | - | 11 - 12 | 1760 | 62 | ||
| AK138760.1 | - | 1779 | 62 | |||
| AK149771.1 | - | 13 - 14 | 1907 | 62 | ||
| AK151300.1 | - | 12 - 13 | 1775 | 62 | ||
| AK152439.1 | - | 12 - 13 | 1775 | 62 | ||
| AK159938.1 | - | 12 - 13 | 1778 | 62 | ||
| AK193271.1 | - | 3 - 4 | 214 | 62 | ||
| BC026573.1 | - | 4 - 5 | 328 | 62 | ||
| BC043100.1 | - | 12 - 13 | 1998 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gtf3c2
Genomic Map