Assay Details
| Gene Symbol: Gtf3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027901.2 | NP_082177.2 | 16 - 17 | 2521 | 144 | |
| GenBank mRNA | AK004895.1 | - | 16 - 17 | 2299 | 144 | |
| AK012095.1 | - | 9 - 10 | 1118 | 144 | ||
| AK083585.1 | - | 16 - 17 | 2523 | 144 | ||
| AK083636.1 | - | 15 - 16 | 2276 | 144 | ||
| AK129033.1 | - | 15 - 16 | 2271 | 144 | ||
| AK138760.1 | - | 2290 | 144 | |||
| AK149771.1 | - | 17 - 18 | 2418 | 144 | ||
| AK151300.1 | - | 16 - 17 | 2286 | 144 | ||
| AK152439.1 | - | 16 - 17 | 2286 | 144 | ||
| AK159938.1 | - | 16 - 17 | 2289 | 144 | ||
| BC026573.1 | - | 8 - 9 | 839 | 144 | ||
| BC034369.1 | - | 16 - 17 | 2299 | 3994677 | 144 | |
| BC043100.1 | - | 15 - 16 | 2687 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gtf3c2
Genomic Map