Assay Details
| Gene Symbol: Gtf3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027901.2 | NP_082177.2 | 17 - 18 | 2684 | 80 | |
| GenBank mRNA | AK012095.1 | - | 10 - 11 | 1281 | 80 | |
| AK083585.1 | - | 17 - 18 | 2686 | 80 | ||
| AK083636.1 | - | 16 - 17 | 2439 | 80 | ||
| AK129033.1 | - | 16 - 17 | 2434 | 80 | ||
| AK138760.1 | - | 2453 | 80 | |||
| AK151300.1 | - | 17 - 18 | 2449 | 80 | ||
| AK152439.1 | - | 17 - 18 | 2449 | 80 | ||
| AK159938.1 | - | 17 - 18 | 2452 | 80 | ||
| BC026573.1 | - | 9 - 10 | 1002 | 80 | ||
| BC034369.1 | - | 17 - 18 | 2462 | 3994677 | 80 | |
| BC043100.1 | - | 16 - 17 | 2850 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gtf3c2
Genomic Map