Assay Details
Gene Symbol: Kat6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081149.1 | NP_001074618.1 | 11 - 12 | 2230 | 103 | |
GenBank mRNA | AK028058.1 | - | 10 - 11 | 2149 | 103 | |
AK036885.1 | - | 11 - 12 | 2225 | 103 | ||
AK082463.1 | - | 11 - 12 | 2233 | 103 | ||
AK132474.1 | - | 10 - 11 | 2138 | 103 | ||
AK143408.1 | - | 11 - 12 | 2221 | 103 | ||
BC024786.1 | - | 3 - 4 | 356 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
