Assay Details
| Gene Symbol: Kat6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081149.1 | NP_001074618.1 | 10 - 11 | 2093 | 150 | |
| GenBank mRNA | AK028058.1 | - | 9 - 10 | 2012 | 150 | |
| AK036885.1 | - | 10 - 11 | 2088 | 150 | ||
| AK082463.1 | - | 10 - 11 | 2096 | 150 | ||
| AK132474.1 | - | 9 - 10 | 2001 | 150 | ||
| AK143408.1 | - | 10 - 11 | 2084 | 150 | ||
| BC024786.1 | - | 2 - 3 | 219 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kat6a
Genomic Map