Assay Details
| Gene Symbol: Agpat6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018743.4 | NP_061213.2 | 6 - 7 | 1660 | 66 | |
| GenBank mRNA | AF015811.1 | - | 3 - 4 | 336 | 66 | |
| AF406611.1 | - | 5 - 6 | 1171 | 66 | ||
| AK045235.1 | - | 6 - 7 | 1554 | 66 | ||
| AK049675.1 | - | 6 - 7 | 1553 | 66 | ||
| AK077561.1 | - | 6 - 7 | 1544 | 66 | ||
| AK083589.1 | - | 6 - 7 | 1554 | 66 | ||
| AK161270.1 | - | 6 - 7 | 1523 | 66 | ||
| AK162033.1 | - | 3 - 4 | 465 | 66 | ||
| AK167876.1 | - | 6 - 7 | 1569 | 66 | ||
| AK169257.1 | - | 6 - 7 | 1554 | 66 | ||
| AY489184.1 | - | 5 - 6 | 729 | 66 | ||
| BC031767.1 | - | 5 - 6 | 1014 | 4457846 | 66 | |
| BC057860.1 | - | 3 - 4 | 334 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agpat6
Genomic Map