Assay Details
| Gene Symbol: Slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113423.2 | NP_001106894.1 | 5 - 6 | 1492 | 75 | |
| NM_153567.3 | NP_705795.2 | 5 - 6 | 1492 | 75 | ||
| GenBank mRNA | AK129365.1 | - | 5 - 6 | 1481 | 75 | |
| AK146432.1 | - | 5 - 6 | 1491 | 75 | ||
| AK216525.1 | - | 1 - 2 | 77 | 75 | ||
| BC019454.1 | - | 3 - 4 | 75 | |||
| BC038019.1 | - | 3 - 4 | 697 | 3597423 | 75 | |
| BC076573.1 | - | 5 - 6 | 1425 | 75 | ||
| CK619668.1 | - | 1 - 2 | 199 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slain2
Genomic Map