Assay Details
| Gene Symbol: Tmprss11e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172880.2 | NP_766468.1 | 4 - 5 | 372 | 135 | |
| GenBank mRNA | AK037235.1 | - | 4 - 5 | 374 | 135 | |
| AK162701.1 | - | 4 - 5 | 369 | 135 | ||
| AY787860.1 | - | 4 - 5 | 380 | 135 | ||
| BC115432.1 | - | 4 - 5 | 366 | 40094164 | 135 | |
| BC115433.1 | - | 4 - 5 | 366 | 40094170 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss11e
Genomic Map