Assay Details
Gene Symbol: Fbxw4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013907.2 | NP_038935.1 | 4 - 5 | 830 | 102 | |
GenBank mRNA | AB022163.1 | - | 4 - 5 | 830 | 102 | |
BC027031.1 | - | 4 - 5 | 692 | 5364017 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
