Assay Details
Gene Symbol: Map2k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042557.2 | NP_001036022.1 | 8 - 9 | 1065 | 83 | |
NM_001164172.1 | NP_001157644.1 | 8 - 9 | 1065 | 83 | ||
NM_001291777.1 | NP_001278706.1 | 7 - 8 | 1017 | 83 | ||
NM_001291778.1 | NP_001278707.1 | 7 - 8 | 908 | 83 | ||
NM_001291783.1 | NP_001278712.1 | 7 - 8 | 908 | 83 | ||
NM_011944.3 | NP_036074.2 | 7 - 8 | 1017 | 83 | ||
GenBank mRNA | AB005654.1 | - | 8 - 9 | 940 | 83 | |
AF022112.1 | - | 7 - 8 | 994 | 83 | ||
AF022113.1 | - | 8 - 9 | 1042 | 83 | ||
AF060943.1 | - | 7 - 8 | 908 | 83 | ||
AF060944.1 | - | 7 - 8 | 928 | 83 | ||
AF060945.1 | - | 7 - 8 | 928 | 83 | ||
AF060946.1 | - | 8 - 9 | 976 | 83 | ||
AF060947.1 | - | 8 - 9 | 976 | 83 | ||
AK031137.1 | - | 7 - 8 | 926 | 83 | ||
AK165184.1 | - | 7 - 8 | 934 | 83 | ||
BC070467.1 | - | 8 - 9 | 977 | 30615302 | 83 | |
U93030.1 | - | 7 - 8 | 908 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
