| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800000203.mm.1 |
NM_001195244.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000203.mm.1 |
NM_001195244.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000203.mm.1 |
TC0800000015.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800000204.mm.1 |
NM_001195244.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000204.mm.1 |
NM_001195244.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000204.mm.1 |
TC0800000015.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800000205.mm.1 |
NM_001195244.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000205.mm.1 |
NM_001195244.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800000205.mm.1 |
TC0800000015.mm.1 |
MTA TC |