Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200045847.mm.1 |
NM_001081092.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045847.mm.1 |
NM_001081092.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045847.mm.1 |
TC0200005397.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200045846.mm.1 |
NM_001081092.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045846.mm.1 |
NM_001081092.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045846.mm.1 |
TC0200005397.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200045852.mm.1 |
NM_001081092.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045852.mm.1 |
NM_001081092.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045852.mm.1 |
TC0200005397.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200045848.mm.1 |
NM_001081092.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045848.mm.1 |
NM_001081092.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200045848.mm.1 |
TC0200005397.mm.1 |
MTA TC |